1-Bromo-2-(2,2-dibromovinyl)-4-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

90040-1G 90040-25G 90040-5G 90040-500MG
76893-876EA 109.28 USD
76893-876 76893-878 76893-880 77652-178
1-Bromo-2-(2,2-dibromovinyl)-4-methoxybenzene ≥95%
1-Bromo-2-(2,2-dibromovinyl)-4-methoxybenzene

Precaución: for Research Use Only

Formula: C₉H₇Br₃O
MW: 370.863 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD34772113
CAS Number: 213599-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR