1-(tert-Butyl)-4-(2,2-dibromovinyl)benzene ≥95%

Supplier: AOBChem USA

Warning

89708-25G 89708-5G 89708-1G 89708-500MG
76893-850EA 587.74 USD
76893-850 76893-852 77644-176 77652-170
1-(tert-Butyl)-4-(2,2-dibromovinyl)benzene ≥95%
1-(tert-Butyl)-4-(2,2-dibromovinyl)benzene

Precaución: for Research Use Only

Formula: C₁₂H₁₄Br₂
MW: 318.048 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD34771979
CAS Number: 1465990-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR