2-(6-Chloro-2-methoxy-3-(methylthio)phenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

89649-1G 89649-25G 89649-500MG 89649-5G
76893-834EA 312.97 USD
76893-834 76893-836 76893-838 76893-840
2-(6-Chloro-2-methoxy-3-(methylthio)phenyl)-1,3-dioxolane ≥97%
2-(6-Chloro-2-methoxy-3-(methylthio)phenyl)-1,3-dioxolane
Formula: C₁₁H₁₃ClO₃S
MW: 260.737 g/mol
Storage Temperature: Refrigerator
CAS Number: 2921831-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR