1,3-Dibromo-2-isopropoxy-5-nitrobenzene ≥97%

Supplier: AOBChem USA

Warning

88003-1G 88003-25G 88003-5G 88003-500MG
76893-800EA 62.6 USD
76893-800 76893-802 76893-804 77652-136
1,3-Dibromo-2-isopropoxy-5-nitrobenzene ≥97%
1,3-Dibromo-2-isopropoxy-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₉H₉Br₂NO₃
Storage Temperature: Ambient
MDL Number: MFCD34563381
CAS Number: 2918942-69-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR