2,4-Dichloro-1-ethylbenzene ≥97%

Supplier: AOBChem USA

Warning

87571-1G 87571-25G 87571-500MG 87571-5G
76893-786EA 184.6 USD
76893-786 76893-788 76893-790 76893-792
2,4-Dichloro-1-ethylbenzene ≥97%
2,4-Dichloro-1-ethylbenzene
Formula: C₈H₈Cl₂
MW: 175.055 g/mol
Storage Temperature: Ambient
CAS Number: 54484-62-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR