1-(4-Fluorophenyl)cyclohexanol ≥95%

Supplier: AOBChem USA

Warning

85020-1G 85020-500MG 85020-5G
76893-734EA 712.93 USD
76893-734 76893-736 76893-738
1-(4-Fluorophenyl)cyclohexanol ≥95%
1-(4-Fluorophenyl)cyclohexanol
Formula: C₁₂H₁₅FO
MW: 194.245 g/mol
Storage Temperature: Refrigerator
CAS Number: 1496-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR