1,4-Dichloro-2,5-dimethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

84224-25G 84224-5G
76893-722EA 516.66 USD
76893-722 76893-724
1,4-Dichloro-2,5-dimethoxybenzene ≥97%
1,4-Dichloro-2,5-dimethoxybenzene
Formula: C₈H₈Cl₂O₂
Storage Temperature: Ambient
CAS Number: 2675-77-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR