1-Bromo-2-(1,1-difluoroethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

84007-1G 84007-25G 84007-5G
76893-710EA 62.6 USD
76893-710 76893-712 76893-714
1-Bromo-2-(1,1-difluoroethyl)benzene ≥97%
1-Bromo-2-(1,1-difluoroethyl)benzene
Formula: C₈H₇BrF₂
Storage Temperature: Ambient
MDL Number: MFCD09996896
CAS Number: 444581-46-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR