1-Bromo-4-(1,1-difluoroethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

83654-25G 83654-5G
76893-702EA 410.57 USD
76893-702 76893-704
1-Bromo-4-(1,1-difluoroethyl)benzene ≥97%
1-Bromo-4-(1,1-difluoroethyl)benzene
Formula: C₈H₇BrF₂
Storage Temperature: Ambient
MDL Number: MFCD11110333
CAS Number: 1000994-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR