1-Bromo-2-chloro-4-fluoro-5-nitrobenzene ≥95% research grade

Supplier: AOBChem USA

Warning

82980-25G 82980-5G
76893-690EA 431.79 USD
76893-690 76893-692
1-Bromo-2-chloro-4-fluoro-5-nitrobenzene ≥95% research grade
1-Bromo-2-chloro-4-fluoro-5-nitrobenzene
Formula: C₆H₂BrClFNO₂
MW: 254.44 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20528320
CAS Number: 111010-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR