1-Bromo-2-(difluoromethoxy)-3-methoxy-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

82320-1G 82320-500MG 82320-5G 82320-25G
76893-676EA 708.69 USD
76893-676 76893-678 76893-680 77656-918
1-Bromo-2-(difluoromethoxy)-3-methoxy-5-methylbenzene ≥97%
1-Bromo-2-(difluoromethoxy)-3-methoxy-5-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₉BrF₂O₂
MW: 267.067 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34769268
CAS Number: 2091467-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR