1-(benzo[d][1,3]dioxol-5-yl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

80948-1G 80948-25G 80948-500MG 80948-5G
76893-660EA 143.23 USD
76893-660 76893-662 76893-664 76893-666
1-(benzo[d][1,3]dioxol-5-yl)-2,2,2-trifluoroethanone ≥95%
1-(benzo[d][1,3]dioxol-5-yl)-2,2,2-trifluoroethanone
Formula: C₉H₅F₃O₃
Storage Temperature: Refrigerator
CAS Number: 102124-73-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR