1,3-Dibromo-5-(Methylsulfonyl)Benzene ≥97%

Supplier: AOBChem USA

Warning

80668-1G 80668-25G 80668-500MG 80668-5G
76893-646EA 134.74 USD
76893-646 76893-648 76893-650 76893-652
1,3-Dibromo-5-(Methylsulfonyl)Benzene ≥97%
1,3-Dibromo-5-(Methylsulfonyl)Benzene
Formula: C₇H₆Br₂O₂S
MW: 313.994 g/mol
Storage Temperature: Ambient
CAS Number: 1222407-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR