2-(5-Bromo-4-fluoro-2-methoxyphenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

80327-1G 80327-25G 80327-500MG 80327-5G
76893-634EA 434.97 USD
76893-634 76893-636 76893-638 76893-640
2-(5-Bromo-4-fluoro-2-methoxyphenyl)-1,3-dioxolane ≥97%
2-(5-Bromo-4-fluoro-2-methoxyphenyl)-1,3-dioxolane
Formula: C₁₀H₁₀BrFO₃
MW: 277.087 g/mol
Storage Temperature: Refrigerator
CAS Number: 2901109-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR