1-Ethyl-5-iodo-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

79997-1G 79997-25G 79997-5G 79997-500MG
76893-620EA 100.79 USD
76893-620 76893-622 76893-624 77648-002
1-Ethyl-5-iodo-1H-pyrazole ≥97%
1-Ethyl-5-iodo-1H-pyrazole
Formula: C₅H₇IN₂
Storage Temperature: Refrigerator
MDL Number: MFCD24448831
CAS Number: 1392274-29-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR