2,4-Dibromo-3,6-difluoroaniline ≥97%

Supplier: AOBChem USA

Warning

78635-25G 78635-5G
76893-564EA 403.15 USD
76893-564 76893-566
2,4-Dibromo-3,6-difluoroaniline ≥97%
2,4-Dibromo-3,6-difluoroaniline

Precaución: for Research Use Only

Formula: C₆H₃Br₂F₂N
Storage Temperature: Ambient
MDL Number: MFCD09878118
CAS Number: 1000577-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR