1-(2-Fluoro-5-methoxyphenyl)ethanone ≥97% research grade

Supplier: AOBChem USA

Warning

78047-1G 78047-25G 78047-500MG 78047-5G
76893-534EA 228.1 USD
76893-534 76893-536 76893-538 76893-540
1-(2-Fluoro-5-methoxyphenyl)ethanone ≥97% research grade
1-(2-Fluoro-5-methoxyphenyl)ethanone
Formula: C₉H₉FO₂
MW: 168.165 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01074363
CAS Number: 80309-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR