1-Bromo-2-(methoxymethoxy)-3-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

77594-1G 77594-25G 77594-500MG 77594-5G
76893-518EA 131.56 USD
76893-518 76893-520 76893-522 76893-524
1-Bromo-2-(methoxymethoxy)-3-(trifluoromethyl)benzene ≥97%
1-Bromo-2-(methoxymethoxy)-3-(trifluoromethyl)benzene
Formula: C₉H₈BrF₃O₂
MW: 285.0578 g/mol
Storage Temperature: Refrigerator
CAS Number: 2918947-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR