(2-Cyano-5-methoxyphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

76892-1G 76892-500MG 76892-5G
76893-506EA 591.99 USD
76893-506 76893-508 76893-510
(2-Cyano-5-methoxyphenyl)boronic acid ≥97%
(2-Cyano-5-methoxyphenyl)boronic acid
Formula: C₈H₈BNO₃
MW: 176.965 g/mol
Storage Temperature: Refrigerator
CAS Number: 1333265-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR