1-(4-Chloro-2-fluorophenyl)ethanol 95%

Supplier: AOBChem USA

Warning

76175-25G 76175-5G 76175-1G
76893-486EA 630.18 USD
76893-486 76893-488 77639-982
1-(4-Chloro-2-fluorophenyl)ethanol 95%
1-(4-Chloro-2-fluorophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₈ClFO
Storage Temperature: Refrigerator
MDL Number: MFCD09037960
CAS Number: 933673-55-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR