2-Fluoro-5-iodo-4-methylanisole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Fluoro-4-iodo-2-methoxy-5-methylbenzene

76060-1G 76060-25G 76060-500MG 76060-5G
76893-470EA 366.02 USD
76893-470 76893-472 76893-474 76893-476
2-Fluoro-5-iodo-4-methylanisole ≥97%
2-Fluoro-5-iodo-4-methylanisole

Precaución: for Research Use Only

Formula: C₈H₈FIO
MW: 266.051 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27975454
CAS Number: 1823867-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR