5-Bromo-2-fluoro-3-(trifluoromethyl)benzyl alcohol ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
(5-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)methanol
75540-1G
75540-25G
75540-500MG
75540-5G
76893-442EA
214.31
USD
76893-442
76893-444
76893-446
76893-448
5-Bromo-2-fluoro-3-(trifluoromethyl)benzyl alcohol ≥97%
5-Bromo-2-fluoro-3-(trifluoromethyl)benzyl alcohol
Precaución: for Research Use Only
Formula:
C₈H₅BrF₄O MW: 273.022 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32640122 CAS Number: 2090690-47-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...