5-Bromo-4-chloro-2-fluorobenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (5-Bromo-4-chloro-2-fluorophenyl)methanol

74843-1G 74843-25G 74843-500MG 74843-5G
76893-414EA 211.12 USD
76893-414 76893-416 76893-418 76893-420
5-Bromo-4-chloro-2-fluorobenzyl alcohol ≥97%
5-Bromo-4-chloro-2-fluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₅BrClFO
MW: 239.469 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28060809
CAS Number: 1785275-10-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR