1-(5-Bromo-3-chloro-2-fluorophenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

74429-1G 74429-500MG 74429-5G
76893-400EA 619.57 USD
76893-400 76893-402 76893-404
1-(5-Bromo-3-chloro-2-fluorophenyl)ethanone ≥97%
1-(5-Bromo-3-chloro-2-fluorophenyl)ethanone
Formula: C₈H₅BrClFO
Storage Temperature: Ambient
MDL Number: MFCD26679371
CAS Number: 1536939-67-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR