(2-Bromo-4-Chloro-3-Fluorophenyl)(Methyl)Sulfane ≥97%

Supplier: AOBChem USA

Warning

74028-1G 74028-500MG 74028-5G 74028-25G
76893-372EA 650.34 USD
76893-372 76893-374 76893-376 77656-858
(2-Bromo-4-Chloro-3-Fluorophenyl)(Methyl)Sulfane ≥97%
(2-Bromo-4-Chloro-3-Fluorophenyl)(Methyl)Sulfane

Precaución: for Research Use Only

Formula: C₇H₅BrClFS
MW: 255.535 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34765266
CAS Number: 2709426-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR