1-tert-Butylpyrazole-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-(tert-Butyl)-1H-pyrazole-4-carbaldehyde

72837-25G 72837-5G
76893-346EA 418 USD
76893-346 76893-348
1-tert-Butylpyrazole-4-carbaldehyde ≥97%
1-tert-Butylpyrazole-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₁₂N₂O
MW: 152.194 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09939615
CAS Number: 127949-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR