1-Chloro-5-fluoro-2-(methoxymethoxy)-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

72745-1G 72745-25G 72745-500MG 72745-5G
76893-330EA 255.68 USD
76893-330 76893-332 76893-334 76893-336
1-Chloro-5-fluoro-2-(methoxymethoxy)-3-methylbenzene ≥97%
1-Chloro-5-fluoro-2-(methoxymethoxy)-3-methylbenzene
Formula: C₉H₁₀ClFO₂
MW: 204.626 g/mol
Storage Temperature: Refrigerator
CAS Number: 2807444-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR