4'-Phenyl-2,2,2-trifluoroacetophenone ≥97%

Supplier: AOBChem USA

Warning

72619-1G 72619-25G 72619-500MG 72619-5G
76893-304EA 280.08 USD
76893-304 76893-306 76893-308 76893-310
4'-Phenyl-2,2,2-trifluoroacetophenone ≥97%
4'-Phenyl-2,2,2-trifluoroacetophenone
Formula: C₁₄H₉F₃O
Storage Temperature: Ambient
MDL Number: MFCD01320014
CAS Number: 2369-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR