1,2-Cichloro-3-isopropoxybenzene 97%

Supplier: AOBChem USA

Warning

72380-1G 72380-25G 72380-5G 72380-500MG
76893-290EA 102.91 USD
76893-290 76893-292 76893-294 77651-970
1,2-Cichloro-3-isopropoxybenzene 97%
1,2-Cichloro-3-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₀Cl₂O
MW: 205.081 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23114217
CAS Number: 197634-45-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR