2,2-Difluoro-1-(4-fluorophenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

71581-1G 71581-25G 71581-500MG 71581-5G
76893-274EA 340.55 USD
76893-274 76893-276 76893-278 76893-280
2,2-Difluoro-1-(4-fluorophenyl)ethanone ≥97%
2,2-Difluoro-1-(4-fluorophenyl)ethanone
Formula: C₈H₅F₃O
MW: 174.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16618056
CAS Number: 50562-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR