(4-Chloro-3-Fluorophenyl)(Methyl)Sulfane ≥97%

Supplier: AOBChem USA

Warning

71409-1G 71409-25G 71409-500MG 71409-5G
76893-260EA 266.29 USD
76893-260 76893-262 76893-264 76893-266
(4-Chloro-3-Fluorophenyl)(Methyl)Sulfane ≥97%
(4-Chloro-3-Fluorophenyl)(Methyl)Sulfane
Formula: C₇H₆ClFS
MW: 176.6389 g/mol
Storage Temperature: Refrigerator
CAS Number: 1314964-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR