(4-Fluoro-3-iodophenyl)methanol ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 4-Fluoro-3-iodobenzyl alcohol

69518-1G 69518-25G 69518-500MG 69518-5G
76893-238EA 169.75 USD
76893-238 76893-240 76893-242 76893-244
(4-Fluoro-3-iodophenyl)methanol ≥97% research grade
(4-Fluoro-3-iodophenyl)methanol
Formula: C₇H₆FIO
Storage Temperature: Ambient
MDL Number: MFCD18396949
CAS Number: 227609-87-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR