1,2-Dibromo-4-fluoro-5-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

69460-25G 69460-5G
76893-226EA 513.48 USD
76893-226 76893-228
1,2-Dibromo-4-fluoro-5-iodobenzene ≥97%
1,2-Dibromo-4-fluoro-5-iodobenzene
Formula: C₆H₂Br₂FI
Storage Temperature: Ambient
CAS Number: 1000578-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR