(5-Chloro-6-Fluoro-[1,1'-Biphenyl]-3-Yl)(Methyl)Sulfane ≥97%

Supplier: AOBChem USA

Warning

68539-1G 68539-500MG 68539-5G 68539-25G
76893-174EA 537.88 USD
76893-174 76893-176 76893-178 77656-814
(5-Chloro-6-Fluoro-[1,1'-Biphenyl]-3-Yl)(Methyl)Sulfane ≥97%
(5-Chloro-6-Fluoro-[1,1'-Biphenyl]-3-Yl)(Methyl)Sulfane

Precaución: for Research Use Only

Formula: C₁₃H₁₀ClFS
MW: 252.735 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763465
CAS Number: 2918946-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR