2,2,2-Trifluoro-1-(2-methyl-4-(trifluoromethyl)phenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

67552-1G 67552-25G 67552-500MG 67552-5G
76893-144EA 147.47 USD
76893-144 76893-146 76893-148 76893-150
2,2,2-Trifluoro-1-(2-methyl-4-(trifluoromethyl)phenyl)ethanone ≥95%
2,2,2-Trifluoro-1-(2-methyl-4-(trifluoromethyl)phenyl)ethanone
Formula: C₁₀H₆F₆O
MW: 256.144 g/mol
Storage Temperature: Ambient
CAS Number: 1881329-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR