2,3,4-Trimethoxybenzamide ≥97%

Supplier: AOBChem USA

Warning

66565-1G 66565-25G 66565-5G 66565-500MG
76893-110EA 79.57 USD
76893-110 76893-112 76893-114 77651-918
2,3,4-Trimethoxybenzamide ≥97%
2,3,4-Trimethoxybenzamide

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₄
MW: 211.215 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09870479
CAS Number: 4304-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR