3-Benzyloxy-5-(trifluoromethoxy)phenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: (3-(Benzyloxy)-5-(trifluoromethoxy)phenyl)boronic acid

66429-1G 66429-25G 66429-5G 66429-500MG
76893-084EA 102.91 USD
76893-084 76893-086 76893-088 77647-878
3-Benzyloxy-5-(trifluoromethoxy)phenylboronic acid ≥97% research grade
3-Benzyloxy-5-(trifluoromethoxy)phenylboronic acid
Formula: C₁₄H₁₂BF₃O₄
Storage Temperature: Ambient
MDL Number: MFCD16036111
CAS Number: 1256345-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR