1-(Benzyloxy)-2-bromo-4-(difluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

66263-1G 66263-500MG 66263-5G 66263-25G
76893-070EA 589.87 USD
76893-070 76893-072 76893-074 77656-788
1-(Benzyloxy)-2-bromo-4-(difluoromethyl)benzene ≥97%
1-(Benzyloxy)-2-bromo-4-(difluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₄H₁₁BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD31735280
CAS Number: 2709493-49-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR