2,2,2-Trifluoro-1-(4-fluoro-3-methylphenyl)ethanol ≥95% research grade

Supplier: AOBChem USA

Warning

65808-1G 65808-25G 65808-500MG 65808-5G
76893-040EA 192.03 USD
76893-040 76893-042 76893-044 76893-046
2,2,2-Trifluoro-1-(4-fluoro-3-methylphenyl)ethanol ≥95% research grade
2,2,2-Trifluoro-1-(4-fluoro-3-methylphenyl)ethanol
Formula: C₉H₈F₄O
Storage Temperature: Ambient
CAS Number: 1251084-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR