2',4'-Dichloro-2,2,2-trifluoroacetophenone ≥97%

Supplier: AOBChem USA

Warning

65669-25G 65669-5G
76893-028EA 198.39 USD
76893-028 76893-030
2',4'-Dichloro-2,2,2-trifluoroacetophenone ≥97%
2',4'-Dichloro-2,2,2-trifluoroacetophenone
Formula: C₈H₃Cl₂F₃O
Storage Temperature: Ambient
MDL Number: MFCD02260844
CAS Number: 92736-81-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR