2',6'-Difluoro-3'-methoxyacetophenone ≥97%

Supplier: AOBChem USA

Warning

65604-1G 65604-25G 65604-5G
76893-016EA 64.72 USD
76893-016 76893-018 76893-020
2',6'-Difluoro-3'-methoxyacetophenone ≥97%
2',6'-Difluoro-3'-methoxyacetophenone

Precaución: for Research Use Only

Formula: C₉H₈F₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD20487626
CAS Number: 1206593-23-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR