2,3,6-Trifluorobenzonitrile ≥97%

Supplier: AOBChem USA

Warning

65423-25G 65423-5G
76893-004EA 190.97 USD
76893-004 76893-006
2,3,6-Trifluorobenzonitrile ≥97%
2,3,6-Trifluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₂F₃N
MW: 157.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012287
CAS Number: 136514-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR