(2,3-Difluorophenyl)trimethylsilane 95%

Supplier: AOBChem USA

Warning

65227-1G 65227-25G 65227-5G 65227-500MG
76892-992EA 114.58 USD
76892-992 76892-994 76892-996 77651-898
(2,3-Difluorophenyl)trimethylsilane 95%
(2,3-Difluorophenyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₉H₁₂F₂Si
MW: 186.2738 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28363370
CAS Number: 138871-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR