(2-Chloro-3-pyridinyl)(4-morpholinyl)methanone ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-(2-Chloropyridine-3-carbonyl)morpholine, (2-Chloropyridin-3-yl)(morpholino)methanone

64847-1G 64847-25G 64847-5G
76892-950EA 62.6 USD
76892-950 76892-952 76892-954
(2-Chloro-3-pyridinyl)(4-morpholinyl)methanone ≥97%
(2-Chloro-3-pyridinyl)(4-morpholinyl)methanone
Formula: C₁₀H₁₁ClN₂O₂
MW: 226.66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00454210
CAS Number: 53062-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR