1-Fluoro-4-isopropylbenzene ≥97%

Supplier: AOBChem USA

Warning

64699-1G 64699-25G 64699-5G
76892-936EA 62.6 USD
76892-936 76892-938 76892-940
1-Fluoro-4-isopropylbenzene ≥97%
1-Fluoro-4-isopropylbenzene
Formula: C₉H₁₁F
MW: 138.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00017969
CAS Number: 403-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR