(4-Bromophenyl)(p-tolyl)methanol ≥97%

Supplier: AOBChem USA

Warning

64455-1G 64455-25G 64455-500MG 64455-5G
76892-920EA 123.07 USD
76892-920 76892-922 76892-924 76892-926
(4-Bromophenyl)(p-tolyl)methanol ≥97%
(4-Bromophenyl)(p-tolyl)methanol
Formula: C₁₄H₁₃BrO
MW: 277.1564 g/mol
Storage Temperature: Ambient
CAS Number: 29334-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR