2,4-Dichloro-6-fluorophenol ≥97%

Supplier: AOBChem USA

Warning

64418-1G 64418-25G 64418-500MG 64418-5G
76892-904EA 206.88 USD
76892-904 76892-906 76892-908 76892-910
2,4-Dichloro-6-fluorophenol ≥97%
2,4-Dichloro-6-fluorophenol
Formula: C₆H₃Cl₂FO
MW: 180.992 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12405440
CAS Number: 344-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR