1-Bromo-4-ethoxynaphthalene ≥95%

Supplier: AOBChem USA

Warning

64372-1G 64372-25G 64372-5G
76892-892EA 79.57 USD
76892-892 76892-894 76892-896
1-Bromo-4-ethoxynaphthalene ≥95%
1-Bromo-4-ethoxynaphthalene
Formula: C₁₂H₁₁BrO
Storage Temperature: Ambient
CAS Number: 20900-22-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR