4'-tert-Butyl-2,2,2-trifluoroacetophenone ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1-(4-(Tert-butyl)phenyl)-2,2,2-trifluoroethanone

64222-1G 64222-25G 64222-5G
76892-878EA 62.6 USD
76892-878 76892-880 76892-882
4'-tert-Butyl-2,2,2-trifluoroacetophenone ≥95%
4'-tert-Butyl-2,2,2-trifluoroacetophenone

Precaución: for Research Use Only

Formula: C₁₂H₁₃F₃O
Storage Temperature: Ambient
MDL Number: MFCD01319979
CAS Number: 73471-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR