4'-Methoxy-2,2,2-trifluoroacetophenone ≥95%

Supplier: AOBChem USA

Warning

63757-25G 63757-5G
76892-848EA 346.92 USD
76892-848 76892-850
4'-Methoxy-2,2,2-trifluoroacetophenone ≥95%
4'-Methoxy-2,2,2-trifluoroacetophenone

Precaución: for Research Use Only

Formula: C₉H₇F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD00018061
CAS Number: 711-38-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR